Product Details

SNP ID
rs28381354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31742835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGGGTTTTCTCTTAGTCAAAGA[C/G]AAAGATCCTTTAACTCATTTGATCT
Phenotype
MIM: 602872 MIM: 603382
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLIC1 PubMed Links

Gene Details

Gene
CLIC1
Gene Name
chloride intracellular channel 1
There are no transcripts associated with this gene.

Gene
MSH5
Gene Name
mutS homolog 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002441.4 Intron NP_002432.1
NM_025259.5 Intron NP_079535.4
NM_172165.3 Intron NP_751897.1
NM_172166.3 Intron NP_751898.1
Gene
MSH5-SAPCD1
Gene Name
MSH5-SAPCD1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details