Product Details

SNP ID
rs35175000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31827898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGCTCTTGTCGCGGATCCCGTC[C/T]GCCGTTTCCAGCCCCCAGTCTCAGA
Phenotype
MIM: 605447 MIM: 140550 MIM: 603012
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C6orf48 PubMed Links

Gene Details

Gene
C6orf48
Gene Name
chromosome 6 open reading frame 48
There are no transcripts associated with this gene.

Gene
HSPA1A
Gene Name
heat shock protein family A (Hsp70) member 1A
There are no transcripts associated with this gene.

Gene
HSPA1B
Gene Name
heat shock protein family A (Hsp70) member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005346.4 164 UTR 5 NP_005337.2
Gene
SNORD48
Gene Name
small nucleolar RNA, C/D box 48
There are no transcripts associated with this gene.

Gene
SNORD52
Gene Name
small nucleolar RNA, C/D box 52
There are no transcripts associated with this gene.

View Full Product Details