Product Details

SNP ID
rs28455809
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.6:32557482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAGATTATATGAAGGGTGTGTG[C/T]CAGGGACAGTTTGGAACTGGCCTCC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HLA-DRB6 PubMed Links

Gene Details

Gene
HLA-DRB6
Gene Name
major histocompatibility complex, class II, DR beta 6 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details