Product Details
- SNP ID
-
rs11963013
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:42226542 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCTGAAATGGCAGCAAATGAAAC[C/T]TTTTTCCCCCTCTAAACAAAAGATG
- Phenotype
-
MIM: 611976
MIM: 610322
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPS10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs575434952] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPS10
- Gene Name
- mitochondrial ribosomal protein S10
There are no transcripts associated with this gene.
- Gene
- TRERF1
- Gene Name
- transcriptional regulating factor 1
View Full Product Details