Product Details

SNP ID
rs28774690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:33908453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATTCTCCCCACAGCATTCCCC[C/T]CTTTTCAGTACTCCTACCCCTGCTA
Phenotype
MIM: 608699
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMPER PubMed Links

Gene Details

Gene
BMPER
Gene Name
BMP binding endothelial regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133468.4 Intron NP_597725.1
XM_005249633.2 Intron XP_005249690.1
XM_017011800.1 Intron XP_016867289.1
XM_017011801.1 Intron XP_016867290.1

View Full Product Details