Product Details

SNP ID
rs28503255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:42923394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAATAAGTAAAGAAACTCCAAA[C/T]GGACGAACACCACTGCAGGGAGGAA
Phenotype
MIM: 611839 MIM: 176842
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL32 PubMed Links

Gene Details

Gene
MRPL32
Gene Name
mitochondrial ribosomal protein L32
There are no transcripts associated with this gene.

Gene
PSMA2
Gene Name
proteasome subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002787.4 435 Silent Mutation CCA,CCG P129P NP_002778.1

View Full Product Details