Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001284330.1 | Intron | NP_001271259.1 | |||
NM_001284331.1 | Intron | NP_001271260.1 | |||
NM_013284.3 | Intron | NP_037416.1 | |||
XM_005249711.2 | Intron | XP_005249768.1 | |||
XM_006715692.2 | Intron | XP_006715755.1 | |||
XM_006715696.2 | Intron | XP_006715759.1 | |||
XM_006715698.3 | Intron | XP_006715761.1 | |||
XM_011515275.1 | Intron | XP_011513577.1 | |||
XM_011515278.1 | Intron | XP_011513580.1 | |||
XM_011515279.1 | Intron | XP_011513581.1 | |||
XM_011515282.2 | Intron | XP_011513584.1 | |||
XM_011515285.1 | Intron | XP_011513587.1 | |||
XM_011515286.2 | Intron | XP_011513588.1 | |||
XM_011515287.2 | Intron | XP_011513589.1 | |||
XM_017011998.1 | Intron | XP_016867487.1 | |||
XM_017011999.1 | Intron | XP_016867488.1 | |||
XM_017012000.1 | Intron | XP_016867489.1 | |||
XM_017012001.1 | Intron | XP_016867490.1 | |||
XM_017012002.1 | Intron | XP_016867491.1 | |||
XM_017012003.1 | Intron | XP_016867492.1 | |||
XM_017012004.1 | Intron | XP_016867493.1 | |||
XM_017012005.1 | Intron | XP_016867494.1 |