Product Details

SNP ID
rs28708645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76303158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGGCGTGCGGTTGAAACGGGGGT[A/C]CCGGGGGCCTGGGGAGTTAAACGTT
Phenotype
MIM: 602195
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSPB1 PubMed Links

Gene Details

Gene
HSPB1
Gene Name
heat shock protein family B (small) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001540.3 Intron NP_001531.1

View Full Product Details