Product Details

SNP ID
rs1804476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:88209352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGAACTCACCTGTAAGAGCCCT[A/C]AGTTTGACGCAGCAGGCGATGTAGT
Phenotype
MIM: 603709 MIM: 182520
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADAM22 PubMed Links

Gene Details

Gene
ADAM22
Gene Name
ADAM metallopeptidase domain 22
There are no transcripts associated with this gene.

Gene
LOC102723885
Gene Name
uncharacterized LOC102723885
There are no transcripts associated with this gene.

Gene
SRI
Gene Name
sorcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256891.1 556 Silent Mutation CTG,CTT L166L NP_001243820.1
NM_001256892.1 556 Silent Mutation CTG,CTT L151L NP_001243821.1
NM_003130.3 556 Silent Mutation CTG,CTT L166L NP_003121.1
NM_198901.1 556 Silent Mutation CTG,CTT L151L NP_944490.1
XM_011516528.1 556 Silent Mutation CTG,CTT L123L XP_011514830.1

View Full Product Details