Product Details

SNP ID
rs17885823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100889680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCACCGCAGGCCGCAATAGGCA[A/G]CCACCGACCGGCACCGACCGAGGAG
Phenotype
MIM: 100740 MIM: 614469 MIM: 611481
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACHE PubMed Links

Gene Details

Gene
ACHE
Gene Name
acetylcholinesterase (Cartwright blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000665.4 39 Intron NP_000656.1
NM_001282449.1 39 Intron NP_001269378.1
NM_001302621.1 39 Intron NP_001289550.1
NM_001302622.1 39 Intron NP_001289551.1
NM_015831.2 39 Intron NP_056646.1
XM_006715995.2 39 Intron XP_006716058.1
XM_011516225.1 39 Intron XP_011514527.1
XM_011516226.1 39 Intron XP_011514528.1
XM_011516228.2 39 Intron XP_011514530.1
XM_011516229.2 39 Intron XP_011514531.1
XM_017012219.1 39 Intron XP_016867708.1
XM_017012220.1 39 Intron XP_016867709.1
Gene
SRRT
Gene Name
serrate, RNA effector molecule
There are no transcripts associated with this gene.

Gene
UFSP1
Gene Name
UFM1 specific peptidase 1 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015072.3 39 UTR 5 NP_001015072.2

View Full Product Details