Product Details

SNP ID
rs28429010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101236769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTGCACACACACCGCCTCCTTA[C/G]AGCGGACGCTCAACCTGCAACTTGC
Phenotype
MIM: 615778 MIM: 609003
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLDN15 PubMed Links

Gene Details

Gene
CLDN15
Gene Name
claudin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185080.1 Intron NP_001172009.1
NM_014343.2 Intron NP_055158.1
Gene
FIS1
Gene Name
fission, mitochondrial 1
There are no transcripts associated with this gene.

View Full Product Details