Product Details

SNP ID
rs3181008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103103472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTTAGGTATCTTGATTCTCCA[T/C]GCTTCAAGACAAAAAAATCTTCAGC
Phenotype
MIM: 611864 MIM: 612334
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
There are no transcripts associated with this gene.

Gene
NAPEPLD
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122838.1 1467 Missense Mutation CAT,CGT H380R NP_001116310.1
NM_198990.4 1467 Missense Mutation CAT,CGT H380R NP_945341.3
XM_005250214.4 1467 Missense Mutation CAT,CGT H380R XP_005250271.2
XM_005250215.2 1467 Missense Mutation CAT,CGT H380R XP_005250272.1
XM_005250218.2 1467 Missense Mutation CAT,CGT H380R XP_005250275.1
XM_011515933.2 1467 Missense Mutation CAT,CGT H380R XP_011514235.1
XM_011515934.2 1467 Missense Mutation CAT,CGT H245R XP_011514236.1
XM_017011854.1 1467 Missense Mutation CAT,CGT H380R XP_016867343.1
XM_017011855.1 1467 Missense Mutation CAT,CGT H380R XP_016867344.1
XM_017011856.1 1467 Intron XP_016867345.1
XM_017011857.1 1467 Intron XP_016867346.1

View Full Product Details