Product Details

SNP ID
rs34325395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107473363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTTTAGCATAATGTCCCACTT[C/G]TTCTAATTTTTAAGTTTGCCAACTG
Phenotype
MIM: 606821 MIM: 601910
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COG5 PubMed Links

Gene Details

Gene
COG5
Gene Name
component of oligomeric golgi complex 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161520.1 Intron NP_001154992.1
NM_006348.3 Intron NP_006339.3
NM_181733.2 Intron NP_859422.2
Gene
GPR22
Gene Name
G protein-coupled receptor 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005295.2 Intron NP_005286.2
XM_011516056.2 Intron XP_011514358.1

View Full Product Details