Product Details

SNP ID
rs28952014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127346384 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGGCTGAACTGTGTACTTTTCT[C/T]AAAGAAAATGTTCAGTTGTCCCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF800 PubMed Links

Gene Details

Gene
ZNF800
Gene Name
zinc finger protein 800
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176814.4 4456 Intron NP_789784.2
XM_005250181.3 4456 Intron XP_005250238.1
XM_006715867.3 4456 UTR 3 XP_006715930.1
XM_006715868.3 4456 UTR 3 XP_006715931.1
XM_006715869.3 4456 UTR 3 XP_006715932.1
XM_011515859.2 4456 UTR 3 XP_011514161.1

View Full Product Details