Product Details
- SNP ID
-
hCV59145320
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:37698391 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCCGAGAAACTTCTGGCCGCCT[A/C]CCCCGGGGCCTCGGGCCTGGGCAGC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF703
PubMed Links
Gene Details
- Gene
- ZNF703
- Gene Name
- zinc finger protein 703
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025069.2 |
1719 |
Missense Mutation |
TAC,TCC |
Y497S |
NP_079345.1 |
View Full Product Details