Product Details
- SNP ID
-
rs11541910
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:35700199 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTCAAGGATTTCTACCTTGCAAG[A/C]CCGAAGCATATCTGCAATAGCACGG
- Phenotype
-
MIM: 186745
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TLN1
PubMed Links
Gene Details
- Gene
- TLN1
- Gene Name
- talin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006289.3 |
7006 |
Missense Mutation |
GCT,TCT |
A2218S |
NP_006280.3 |
View Full Product Details