Product Details
- SNP ID
-
rs7846863
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:112693451 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTACCTTCTCAGTTTCCCACCC[C/T]AAGAAGCCTAAAATCCCTTTTGTTT
- Phenotype
-
MIM: 613273
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
INIP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75532072] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INIP
- Gene Name
- INTS3 and NABP interacting protein
There are no transcripts associated with this gene.
View Full Product Details