Product Details

SNP ID
rs35600008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128459697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGTAGCAGCCCTCTTTGGTCACC[C/T]TGCTAAAAATGCTTTTGCACACCGT
Phenotype
MIM: 602015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ODF2 PubMed Links

Gene Details

Gene
ODF2
Gene Name
outer dense fiber of sperm tails 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242352.1 Intron NP_001229281.1
NM_001242353.1 Intron NP_001229282.1
NM_001242354.1 Intron NP_001229283.1
NM_002540.4 Intron NP_002531.3
NM_153432.1 Intron NP_702910.1
NM_153433.1 Intron NP_702911.1
NM_153435.1 Intron NP_702913.1
NM_153436.1 Intron NP_702914.1
NM_153437.2 Intron NP_702915.1
NM_153439.1 Intron NP_702917.1
NM_153440.1 Intron NP_702918.1
XM_005252010.2 Intron XP_005252067.1
XM_005252011.2 Intron XP_005252068.1
XM_005252012.3 Intron XP_005252069.1
XM_005252013.3 Intron XP_005252070.1
XM_005252014.3 Intron XP_005252071.1
XM_005252017.4 Intron XP_005252074.1
XM_005252018.2 Intron XP_005252075.1
XM_005252019.3 Intron XP_005252076.1
XM_005252022.2 Intron XP_005252079.1
XM_005252023.2 Intron XP_005252080.1
XM_005252024.3 Intron XP_005252081.1
XM_005252026.4 Intron XP_005252083.1
XM_005252029.2 Intron XP_005252086.1
XM_006717126.3 Intron XP_006717189.1
XM_011518722.2 Intron XP_011517024.1
XM_011518723.2 Intron XP_011517025.1
XM_011518724.2 Intron XP_011517026.1
XM_011518725.2 Intron XP_011517027.1
XM_011518726.2 Intron XP_011517028.1
XM_011518728.2 Intron XP_011517030.1
XM_011518729.2 Intron XP_011517031.1
XM_011518730.2 Intron XP_011517032.1
XM_011518731.2 Intron XP_011517033.1
XM_011518732.2 Intron XP_011517034.1
XM_011518733.2 Intron XP_011517035.1
XM_011518744.1 Intron XP_011517046.1
XM_017014764.1 Intron XP_016870253.1
XM_017014765.1 Intron XP_016870254.1
XM_017014766.1 Intron XP_016870255.1
XM_017014767.1 Intron XP_016870256.1
XM_017014768.1 Intron XP_016870257.1
XM_017014769.1 Intron XP_016870258.1
XM_017014770.1 Intron XP_016870259.1
XM_017014771.1 Intron XP_016870260.1
XM_017014772.1 Intron XP_016870261.1
XM_017014773.1 Intron XP_016870262.1
XM_017014774.1 Intron XP_016870263.1
XM_017014775.1 Intron XP_016870264.1
XM_017014776.1 Intron XP_016870265.1
XM_017014777.1 Intron XP_016870266.1
XM_017014778.1 Intron XP_016870267.1
XM_017014779.1 Intron XP_016870268.1
XM_017014780.1 Intron XP_016870269.1
XM_017014781.1 Intron XP_016870270.1

View Full Product Details