Product Details
- SNP ID
-
rs17455322
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128957801 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTTCTAGCCTGCCCAGCCCCTCA[A/T]TTTACATAAGAGGATTGAGGCCAGA
- Phenotype
-
MIM: 615587
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NUP188
PubMed Links
Gene Details
- Gene
- NUP188
- Gene Name
- nucleoporin 188
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015354.2 |
|
Intron |
|
|
NP_056169.1 |
View Full Product Details