Product Details

SNP ID
rs2479230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96971600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAAAGTCTTATGTAAGGCTAAC[A/G]GGCTAACAGCCACCCTCTAAAGTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFSD14C PubMed Links

Gene Details

Gene
MFSD14C
Gene Name
major facilitator superfamily domain containing 14C
There are no transcripts associated with this gene.

View Full Product Details