Product Details

SNP ID
rs35151751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2380192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACAGGCAGGATGTGCAGTGGG[A/G]AAGGGGCTGCGGGAACCCTGCAGGG
Phenotype
MIM: 186845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD81 PubMed Links

Gene Details

Gene
CD81
Gene Name
CD81 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297649.1 Intron NP_001284578.1
NM_004356.3 Intron NP_004347.1
XM_011520492.2 Intron XP_011518794.1
XM_017018598.1 Intron XP_016874087.1
Gene
CD81-AS1
Gene Name
CD81 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details