Product Details
- SNP ID
-
rs28692753
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:18081579 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAAAGGATTATAAATCAAAAAAG[A/G]AAAAATATTGTGTGTATGTTACTTA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RERGL
PubMed Links
Gene Details
- Gene
- RERGL
- Gene Name
- RERG like
View Full Product Details