Product Details
- SNP ID
-
hCV59957378
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6917604 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCAAGTTTGGGGCCAATGCCATC[A/C]TGGGTGTGTCTCTGGCCGTGTGTAA
- Phenotype
-
MIM: 607462
MIM: 131360
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATN1
PubMed Links
Gene Details
- Gene
- ATN1
- Gene Name
- atrophin 1
There are no transcripts associated with this gene.
- Gene
- ENO2
- Gene Name
- enolase 2
There are no transcripts associated with this gene.
- Gene
- LRRC23
- Gene Name
- leucine rich repeat containing 23
There are no transcripts associated with this gene.
View Full Product Details