Product Details
- SNP ID
-
rs12819334
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:110436719 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATATATATATATATATACACACA[C/T]ACACACACACACACACACACACACA
- Phenotype
-
MIM: 604225
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARPC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs575927245] are located under a probe and SNP(s) [rs73206893] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARPC3
- Gene Name
- actin related protein 2/3 complex subunit 3
View Full Product Details