Product Details
- SNP ID
-
rs28532071
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:47193981 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTGAAACAAGGATAATGTCTGTT[G/T]TGATTACATTTGTTCCCTTAATACT
- Phenotype
-
MIM: 609295
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SEMA6D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201559354] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEMA6D
- Gene Name
- semaphorin 6D
View Full Product Details