Product Details

SNP ID
rs28447539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48191821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCCAACACTTAAGTCTACTGGC[C/T]GGACTTCATCTCCATGGCAACAAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTXN2 PubMed Links

Gene Details

Gene
CTXN2
Gene Name
cortexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145668.1 152 UTR 5 NP_001139140.1
XM_005254378.3 152 Intron XP_005254435.1
XM_006720504.2 152 Intron XP_006720567.1
XM_011521553.1 152 Intron XP_011519855.1
XM_017022178.1 152 UTR 5 XP_016877667.1

View Full Product Details