Product Details

SNP ID
rs28662993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50281752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCACAAAGCCAAAAATATTTGG[C/T]ACCTGGCCATTTATGAAAAAAAACT
Phenotype
MIM: 600610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABPB1 PubMed Links

Gene Details

Gene
GABPB1
Gene Name
GA binding protein transcription factor beta subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320910.1 Intron NP_001307839.1
NM_001320915.1 Intron NP_001307844.1
NM_002041.4 Intron NP_002032.2
NM_005254.5 Intron NP_005245.2
NM_016654.4 Intron NP_057738.1
NM_016655.4 Intron NP_057739.1
NM_181427.3 Intron NP_852092.1
XM_005254274.3 Intron XP_005254331.1
XM_011521426.2 Intron XP_011519728.1
XM_017022053.1 Intron XP_016877542.1
XM_017022054.1 Intron XP_016877543.1

View Full Product Details