Product Details

SNP ID
rs28743026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52307662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAAATTTTCTATGCAGATCAAA[A/G]TAGAATTCCTTTTCCAGACCCAACA
Phenotype
MIM: 160777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO5A PubMed Links

Gene Details

Gene
MYO5A
Gene Name
myosin VA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000259.3 11765 UTR 3 NP_000250.3
NM_001142495.1 11765 UTR 3 NP_001135967.1
XM_005254397.3 11765 Intron XP_005254454.1
XM_011521606.2 11765 UTR 3 XP_011519908.2
XM_011521607.2 11765 Intron XP_011519909.2
XM_011521608.2 11765 Intron XP_011519910.2
XM_011521609.2 11765 Intron XP_011519911.2
XM_011521610.2 11765 Intron XP_011519912.2
XM_011521611.2 11765 Intron XP_011519913.2
XM_011521612.2 11765 Intron XP_011519914.2
XM_017022227.1 11765 Intron XP_016877716.1

View Full Product Details