Product Details

SNP ID
rs28507982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72125369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTATTTTAACTTACATTTATTT[C/G]ATTACTACTCTGATGTCCAGTGTCT
Phenotype
MIM: 604875 MIM: 608659
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYO9A PubMed Links

Gene Details

Gene
MYO9A
Gene Name
myosin IXA
There are no transcripts associated with this gene.

Gene
SENP8
Gene Name
SUMO/sentrin peptidase family member, NEDD8 specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166340.1 Intron NP_001159812.1
NM_001172109.1 Intron NP_001165580.1
NM_001172110.1 Intron NP_001165581.1
NM_001172111.1 Intron NP_001165582.1
NM_145204.3 Intron NP_660205.3
XM_005254157.3 Intron XP_005254214.1
XM_011521215.2 Intron XP_011519517.1
XM_011521217.2 Intron XP_011519519.1

View Full Product Details