Product Details
- SNP ID
-
rs35963373
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:74783467 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCACTTCCCCGCCCATTTTGTA[C/T]ACCCGGGAACCGAGGCTCAAAGGGA
- Phenotype
-
MIM: 124095
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CSK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140161971] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CSK
- Gene Name
- c-src tyrosine kinase
- Gene
- MIR4513
- Gene Name
- microRNA 4513
There are no transcripts associated with this gene.
View Full Product Details