Product Details

SNP ID
rs28493801
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74847653 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGAGGGAGGGGACCAAAATAGC[A/G]CCGCCCTTCCAGGCTCAGGCTGTCA
Phenotype
MIM: 606912 MIM: 613472
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MIR6882 PubMed Links
Additional Information
For this assay, SNP(s) [rs79990621] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MIR6882
Gene Name
microRNA 6882
There are no transcripts associated with this gene.

Gene
SCAMP2
Gene Name
secretory carrier membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320778.1 Intron NP_001307707.1
NM_005697.4 Intron NP_005688.2
XM_017021856.1 Intron XP_016877345.1
Gene
ULK3
Gene Name
unc-51 like kinase 3
There are no transcripts associated with this gene.

View Full Product Details