Product Details

SNP ID
rs28677077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28537710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGGTGGTAAAAGGGACAGAGA[C/T]CTGGGTTCTAATTCTGCCTCTCCCA
Phenotype
MIM: 614812
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUPR1 PubMed Links

Gene Details

Gene
NUPR1
Gene Name
nuclear protein 1, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042483.1 Intron NP_001035948.1
NM_012385.2 Intron NP_036517.1

View Full Product Details