Product Details

SNP ID
rs35278728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28596569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGACTCAGCAAAAGGAGGATCC[C/T]GGGCAGGGTGGCTCCCACCTGTAAT
Phenotype
MIM: 613374 MIM: 171150 MIM: 601292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGF29 PubMed Links

Gene Details

Gene
SGF29
Gene Name
SAGA complex associated factor 29
There are no transcripts associated with this gene.

Gene
SULT1A1
Gene Name
sulfotransferase family 1A member 1
There are no transcripts associated with this gene.

Gene
SULT1A2
Gene Name
sulfotransferase family 1A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001054.3 76 Intron NP_001045.1
NM_177528.2 76 Intron NP_803564.1
XM_006721077.3 76 UTR 5 XP_006721140.1
XM_017023600.1 76 Intron XP_016879089.1
XM_017023601.1 76 UTR 5 XP_016879090.1
XM_017023602.1 76 Intron XP_016879091.1
XM_017023603.1 76 Intron XP_016879092.1

View Full Product Details