Product Details

SNP ID
rs28647979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29401192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCTGAGCACTCTTGAACCTGTG[A/C]AATCTGAAAGGAACAGGAGACTTGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NPIPB11 PubMed Links

Gene Details

Gene
NPIPB11
Gene Name
nuclear pore complex interacting protein family member B11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310137.1 Intron NP_001297066.1

View Full Product Details