Product Details

SNP ID
rs36037011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68737599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGGGTCCTGAGGAGCGGAGCGGC[C/G]TGGAAGCCTCGCGCGCTCCGGACCC
Phenotype
MIM: 192090
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDH1 PubMed Links

Gene Details

Gene
CDH1
Gene Name
cadherin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317184.1 Intron NP_001304113.1
NM_001317185.1 Intron NP_001304114.1
NM_001317186.1 Intron NP_001304115.1
NM_004360.4 Intron NP_004351.1
XM_011523488.1 Intron XP_011521790.1
XM_011523489.1 Intron XP_011521791.1

View Full Product Details