Product Details

SNP ID
rs28470573
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78235993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAATGGTGCTAACCCATTCATG[A/G]GAAGCCACTGCCATGATCCAGTCAT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TMEM235 PubMed Links

Gene Details

Gene
TMEM235
Gene Name
transmembrane protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204210.1 Intron NP_001191139.1
NM_001204211.1 Intron NP_001191140.1
NM_001204212.1 Intron NP_001191141.1
XM_011524620.2 Intron XP_011522922.1
XM_011524624.2 Intron XP_011522926.1
XM_017024459.1 Intron XP_016879948.1
XM_017024460.1 Intron XP_016879949.1
XM_017024461.1 Intron XP_016879950.1

View Full Product Details