Product Details
- SNP ID
-
rs28550346
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:19243132 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTGTTTCTCAAACTCCTGTGTGC[A/G]GCAGAAGCACCCAGAGGGCTTGCTG
- Phenotype
-
MIM: 607263
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EPN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117339802] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EPN2
- Gene Name
- epsin 2
View Full Product Details