Product Details

SNP ID
rs34285921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7687680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCAGAGTCAGGATTCTCGCCGA[C/G]CTGGTGCCGTAGATACTAACATTTT
Phenotype
MIM: 191170 MIM: 612661
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TP53 PubMed Links

Gene Details

Gene
TP53
Gene Name
tumor protein p53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000546.5 109 Intron NP_000537.3
NM_001126112.2 109 Intron NP_001119584.1
NM_001126113.2 109 Intron NP_001119585.1
NM_001126114.2 109 Intron NP_001119586.1
NM_001126115.1 109 Intron NP_001119587.1
NM_001126116.1 109 Intron NP_001119588.1
NM_001126117.1 109 Intron NP_001119589.1
NM_001126118.1 109 Intron NP_001119590.1
NM_001276695.1 109 Intron NP_001263624.1
NM_001276696.1 109 Intron NP_001263625.1
NM_001276697.1 109 Intron NP_001263626.1
NM_001276698.1 109 Intron NP_001263627.1
NM_001276699.1 109 Intron NP_001263628.1
NM_001276760.1 109 Intron NP_001263689.1
NM_001276761.1 109 Intron NP_001263690.1
Gene
WRAP53
Gene Name
WD repeat containing antisense to TP53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143990.1 109 Intron NP_001137462.1
NM_001143991.1 109 Intron NP_001137463.1
NM_001143992.1 109 Intron NP_001137464.1
NM_018081.2 109 Intron NP_060551.2
XM_011523952.2 109 Intron XP_011522254.1
XM_017024812.1 109 UTR 5 XP_016880301.1

View Full Product Details