Product Details
- SNP ID
-
rs28404811
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:45442185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGGAGACATGTCCCAGGTCTGCC[A/G]GAGAGAACCCTGCCAGGCAGCTGCC
- Phenotype
-
MIM: 610591
MIM: 611466
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGAP27
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117649063] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGAP27
- Gene Name
- Rho GTPase activating protein 27
There are no transcripts associated with this gene.
- Gene
- PLEKHM1
- Gene Name
- pleckstrin homology and RUN domain containing M1
View Full Product Details