Product Details

SNP ID
rs28367321
Assay Type
Functionally Tested
NCBI dbSNP Submissions
22
Location
Chr.17:65138970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCTCCCCAGCCACCCCTCCT[C/T]CACCCCAGCATCCCCTCCTCCACCC
Phenotype
MIM: 604067
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RGS9 PubMed Links

Gene Details

Gene
RGS9
Gene Name
regulator of G-protein signaling 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081955.2 Intron NP_001075424.1
NM_001165933.1 Intron NP_001159405.1
NM_003835.3 Intron NP_003826.2
XM_011525426.2 Intron XP_011523728.1

View Full Product Details