Product Details

SNP ID
rs28680239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:65139205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCTCCTCCCCAGCCTTCTC[C/T]TCCTCCATCCTAACTTACCTCTCCT
Phenotype
MIM: 604067
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS9 PubMed Links

Gene Details

Gene
RGS9
Gene Name
regulator of G-protein signaling 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081955.2 Intron NP_001075424.1
NM_001165933.1 Intron NP_001159405.1
NM_003835.3 Intron NP_003826.2
XM_011525426.2 Intron XP_011523728.1

View Full Product Details