Product Details

SNP ID
rs28394251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50548477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGAGCACCTCTCCTGGGGCCCT[A/G]CCTGGAATCGCTGGGGTCCTGGGCC
Phenotype
MIM: 607264 MIM: 613939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNA1G-AS1 PubMed Links

Gene Details

Gene
CACNA1G-AS1
Gene Name
CACNA1G antisense RNA 1
There are no transcripts associated with this gene.

Gene
EPN3
Gene Name
epsin 3
There are no transcripts associated with this gene.

Gene
LOC105371824
Gene Name
uncharacterized LOC105371824
There are no transcripts associated with this gene.

Gene
SPATA20
Gene Name
spermatogenesis associated 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258372.1 Intron NP_001245301.1
NM_001258373.1 Intron NP_001245302.1
NM_022827.3 Intron NP_073738.2

View Full Product Details