Product Details

SNP ID
rs35326132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28746754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCAGGGTAAGGTGGCCGCAAG[C/T]TGACCTTTGCCCTGGTTCTTGGCAG
Phenotype
MIM: 609799 MIM: 602464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM222B PubMed Links

Gene Details

Gene
FAM222B
Gene Name
family with sequence similarity 222 member B
There are no transcripts associated with this gene.

Gene
NEK8
Gene Name
NIMA related kinase 8
There are no transcripts associated with this gene.

Gene
TRAF4
Gene Name
TNF receptor associated factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004295.3 Intron NP_004286.2
XM_011525504.2 Intron XP_011523806.1
XM_011525505.2 Intron XP_011523807.1
XM_011525506.2 Intron XP_011523808.1
XM_011525507.2 Intron XP_011523809.1
XM_011525508.2 Intron XP_011523810.1

View Full Product Details