Product Details

SNP ID
rs11557694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32331641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTAAATTGGTAAAATACAAAAA[A/G]AAAATCTGGATTGAGTTTCAAATCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf75 PubMed Links

Gene Details

Gene
C17orf75
Gene Name
chromosome 17 open reading frame 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022344.3 1401 UTR 3 NP_071739.2
XM_005258022.3 1401 UTR 3 XP_005258079.1
XM_017024940.1 1401 UTR 3 XP_016880429.1
XM_017024941.1 1401 UTR 3 XP_016880430.1
Gene
RHBDL3
Gene Name
rhomboid like 3
There are no transcripts associated with this gene.

View Full Product Details