Product Details

SNP ID
hCV60601873
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12328963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATGTGTGTTCAGAAAAGTACAG[G/T]GTTGACATTTTATTTTTTATGTAAA
Phenotype
MIM: 604581 MIM: 615103
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
AFG3L2 PubMed Links

Gene Details

Gene
AFG3L2
Gene Name
AFG3 like matrix AAA peptidase subunit 2
There are no transcripts associated with this gene.

Gene
TUBB6
Gene Name
tubulin beta 6 class V
There are no transcripts associated with this gene.

View Full Product Details