Product Details

SNP ID
rs17445462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:27937822 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGAAAGATTTTATGCAGCTCCA[A/G]ATAGTCAGAAAATCATCCGCTATGA
Phenotype
MIM: 114020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH2 PubMed Links

Gene Details

Gene
CDH2
Gene Name
cadherin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308176.1 Intron NP_001295105.1
NM_001792.4 Intron NP_001783.2
XM_011525788.1 Intron XP_011524090.1
XM_017025514.1 Intron XP_016881003.1

View Full Product Details