Product Details

SNP ID
rs17881574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38904457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGAGAACCTCCTGCAGTTCCCC[A/T]GCAATCTCTCCTTTGGCAACTCTGT
Phenotype
MIM: 604495 MIM: 604480
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCER2 PubMed Links

Gene Details

Gene
CCER2
Gene Name
coiled-coil glutamate rich protein 2
There are no transcripts associated with this gene.

Gene
NFKBIB
Gene Name
NFKB inhibitor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243116.1 Intron NP_001230045.1
NM_002503.4 Intron NP_002494.2
XM_006723226.2 Intron XP_006723289.2
XM_006723227.3 Intron XP_006723290.3
Gene
SIRT2
Gene Name
sirtuin 2
There are no transcripts associated with this gene.

View Full Product Details