Product Details

SNP ID
rs34306641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42428547 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGAAACAATTTTGATCTGTGAC[A/G]TTTAAAAATATTTATTGACTTCTGT
Phenotype
MIM: 611387 MIM: 151750
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXCL17 PubMed Links

Gene Details

Gene
CXCL17
Gene Name
C-X-C motif chemokine ligand 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198477.2 913 UTR 3 NP_940879.1
Gene
LIPE
Gene Name
lipase E, hormone sensitive type
There are no transcripts associated with this gene.

Gene
LIPE-AS1
Gene Name
LIPE antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details