Product Details

SNP ID
rs28696827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52058069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAAACAAGATTAATGTTCCCT[C/T]TGAAGAATAAACATACATAGTGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF432 PubMed Links

Gene Details

Gene
ZNF432
Gene Name
zinc finger protein 432
There are no transcripts associated with this gene.

Gene
ZNF841
Gene Name
zinc finger protein 841
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136499.1 Intron NP_001129971.1
NM_001321349.1 Intron NP_001308278.1
XM_011526833.2 Intron XP_011525135.1
XM_017026652.1 Intron XP_016882141.1
XM_017026653.1 Intron XP_016882142.1
XM_017026654.1 Intron XP_016882143.1
XM_017026655.1 Intron XP_016882144.1
XM_017026656.1 Intron XP_016882145.1
XM_017026657.1 Intron XP_016882146.1
XM_017026658.1 Intron XP_016882147.1
XM_017026659.1 Intron XP_016882148.1
XM_017026660.1 Intron XP_016882149.1
XM_017026661.1 Intron XP_016882150.1
XM_017026662.1 Intron XP_016882151.1
XM_017026663.1 Intron XP_016882152.1

View Full Product Details