Product Details

SNP ID
rs28418599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35226832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTGAGTTAGACAGTGGCCCCC[A/G]CAAAATGCATGTCCTTCCCAGAACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM187B PubMed Links

Gene Details

Gene
FAM187B
Gene Name
family with sequence similarity 187 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152481.1 Intron NP_689694.1

View Full Product Details